5-Bromo-2-methyl-3-(trifluoromethyl)pyridine 95%, Ambeed.Inc

Supplier: Ambeed
Warning

A243990-250MG A243990-10G
103811-560EA 31 USD
103811-560 103851-384 77013-830 77683-733
5-Bromo-2-methyl-3-(trifluoromethyl)pyridine 95%, Ambeed.Inc
5-Bromo-2-methyl-3-(trifluoromethyl)pyridine

Precaución: For research use only.

Formula: C₇H₅BrF₃N
Storage Temperature: Refrigerator
MDL Number: MFCD18257687
CAS Number: 1211526-51-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR