5-Bromo-2-methylbenzimidazole 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-Bromo-2-methyl-1H-benzimidazole, 5-Bromo-2-methyl-1H-benzo[d]imidazole
A178319-10G
A178319-250MG
A178319-1G
A178319-25G
A178319-5G
103807-318EA
6
USD
103807-318
103845-658
76769-474
76769-476
77083-072
77083-074
77083-076
5-Bromo-2-methylbenzimidazole 97%
5-Bromo-2-methylbenzimidazole
Precaución: for Research Use Only
Formula:
C₈H₇BrN₂ MW: 211.0600 g/mol Boiling Pt: 397.3 °C (760 mm Hg) Storage Temperature: Ambient |
MDL Number:
MFCD00457521 CAS Number: 1964-77-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...