5-Bromo-2-methylbenzoxazole 97%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-2-methyl-1,3-benzoxazole

A272660-100MG A272660-250MG A272660-25G A272660-5G
103813-106EA 21 USD
103813-106 103853-018 76772-162 76772-164 77092-802 77092-804
5-Bromo-2-methylbenzoxazole 97%
5-Bromo-2-methylbenzoxazole

Precaución: for Research Use Only

Formula: C₈H₆BrNO
MW: 212.0400 g/mol
Melting Pt: 61…65 °C
Storage Temperature: Ambient
MDL Number: MFCD08702771
CAS Number: 5676-56-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR