5-Bromo-2-methoxypyridine 98%

Supplier: Ambeed
Warning

A109888-100G A109888-10G A109888-25G A109888-500G A109888-5G
103823-392EA 5 USD
103823-392 77270-802 77270-804 77270-806 77270-808 77283-904
5-Bromo-2-methoxypyridine 98%
5-Bromo-2-methoxypyridine

Precaución: for Research Use Only

Formula: C₆H₆BrNO
MW: 188.02 g/mol
Boiling Pt: 80 °C (12 mm Hg)
Flash Pt: 96 °C
Storage Temperature: Ambient
MDL Number: MFCD01318952
CAS Number: 13472-85-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR