3-Amino-5-bromo-2-methoxypyridine 98%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-2-methoxypyridin-3-amine

A108228-100G A108228-10G A108228-25G A108228-1G A108228-5G
103797-312EA 9 USD
103797-312 103823-090 76982-560 76982-562 76982-564 77309-932 77309-934
3-Amino-5-bromo-2-methoxypyridine 98%
3-Amino-5-bromo-2-methoxypyridine

Precaución: for Research Use Only

Formula: C₆H₇BrN₂O
MW: 203.04 g/mol
Flash Pt: 115 °C
Storage Temperature: Ambient
MDL Number: MFCD07368887
CAS Number: 884495-39-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR