5-Bromo-o-anisidine 98%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-2-anisidine, 5-Bromo-2-methoxyaniline, 2-Amino-4-bromoanisole

A149517-100G A149517-10G A149517-25G A149517-5G
103804-056EA 6 USD
103804-056 103842-172 76768-100 76768-102 77317-678 77317-680
5-Bromo-o-anisidine 98%
5-Bromo-o-anisidine

Precaución: for Research Use Only

Formula: C₇H₈BrNO
MW: 202.05 g/mol
Melting Pt: 94…98 °C
Flash Pt: 120 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04037882
CAS Number: 6358-77-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR