5-Bromo-2-methoxy-4-methyl-3-nitropyridine 97%

Supplier: Ambeed
Warning

A389947-1G A389947-25G A389947-5G A389947-100G A389947-10G A389947-250MG A389947-500G
103826-856EA 11 USD
103826-856 103837-898 76841-272 76841-274 76841-276 77142-708 77142-710 77142-712 77142-714
5-Bromo-2-methoxy-4-methyl-3-nitropyridine 97%
5-Bromo-2-methoxy-4-methyl-3-nitropyridine

Precaución: for Research Use Only

Formula: C₇H₇BrN₂O₃
MW: 247.0500 g/mol
Melting Pt: 88…90 °C
Flash Pt: 136.9 °C
Storage Temperature: Ambient
MDL Number: MFCD06659504
CAS Number: 884495-14-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR