5-Bromo-2-methox-3-methylypyridine 97%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-2-methoxy-3-methylpyridine

A514770-1G A514770-25G A514770-5G A514770-100G A514770-10G
103828-444EA 9 USD
103828-444 103839-546 76843-532 76843-534 76843-536 77017-350 77017-352
5-Bromo-2-methox-3-methylypyridine 97%
5-Bromo-2-methox-3-methylypyridine

Precaución: for Research Use Only

Formula: C₇H₈BrNO
MW: 202.05 g/mol
Boiling Pt: 217.8 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD07375124
CAS Number: 760207-87-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR