5-Bromo-2-methox-3-(trifluoromethyl)ypyridine 95%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-2-methoxy-3-(trifluoromethyl)pyridine

A112221-100G A112221-10G A112221-25G A112221-1G A112221-5G
103797-982EA 7 USD
103797-982 103823-816 77025-816 77025-818 77025-820 77310-618 77310-620
5-Bromo-2-methox-3-(trifluoromethyl)ypyridine 95%
5-Bromo-2-methox-3-(trifluoromethyl)ypyridine

Precaución: for Research Use Only

Formula: C₇H₅BrF₃NO
Boiling Pt: 208 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD13185815
CAS Number: 1214377-42-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR