5-Bromo-2-iodopyrimidine 98%

Supplier: Ambeed
Warning

A112868-100G A112868-10G A112868-25G A112868-1G A112868-5G
103798-122EA 7 USD
103798-122 103823-958 76810-008 76810-010 76810-012 77297-214 77310-788
5-Bromo-2-iodopyrimidine 98%
5-Bromo-2-iodopyrimidine

Precaución: for Research Use Only

Formula: C₄H₂BrIN₂
MW: 284.88 g/mol
Boiling Pt: 341 °C (760 mm Hg)
Melting Pt: 99…103 °C
Flash Pt: 160 °C
Storage Temperature: Ambient
MDL Number: MFCD01318111
CAS Number: 183438-24-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR