5-Bromo-2-iodopyridine 98%

Supplier: Ambeed
Warning

A262156-5G A262156-100G A262156-10G A262156-1G A262156-500G
103852-450EA 14 USD
103852-450 77092-066 77276-564 77276-566 77276-568 77276-570
5-Bromo-2-iodopyridine 98%
5-Bromo-2-iodopyridine

Precaución: for Research Use Only

Formula: C₅H₃BrIN
MW: 283.89 g/mol
Boiling Pt: 258.8 °C (760 mm Hg)
Melting Pt: 113…117 °C
Storage Temperature: Ambient
MDL Number: MFCD00691506
CAS Number: 223463-13-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR