3-Amino-5-bromo-2-iodopyridine 98%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-2-iodopyridin-3-amine

A115834-250MG A115834-1G A115834-5G A115834-100MG
103798-632EA 88 USD
103798-632 103824-492 76982-960 77311-318 77311-320 77889-795
3-Amino-5-bromo-2-iodopyridine 98%
3-Amino-5-bromo-2-iodopyridine

Precaución: for Research Use Only

Formula: C₅H₄BrIN₂
Storage Temperature: Refrigerator
MDL Number: MFCD17014978
CAS Number: 1180678-40-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR