5-Bromo-2-iodophenol 98%

Supplier: Ambeed
Warning

A267093-10G A267093-25G A267093-5G
103812-836EA 16 USD
103812-836 103852-738 76874-970 77092-462 77092-464
5-Bromo-2-iodophenol 98%
5-Bromo-2-iodophenol

Precaución: for Research Use Only

Formula: C₆H₄BrIO
MW: 298.904 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16999931
CAS Number: 858855-11-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR