5-Bromo-2-iodobenzotrifluoride 96%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-2-iodobenzotrifluoride, 1-Bromo-4-iodo-3-(trifluoromethyl)benzene

A157745-100G A157745-10G A157745-1G A157745-5G A157745-25G
103805-040EA 8 USD
103805-040 103843-244 76768-442 76768-444 77300-288 77300-290 77319-082
5-Bromo-2-iodobenzotrifluoride 96%
5-Bromo-2-iodobenzotrifluoride

Precaución: for Research Use Only

Formula: C₇H₃BrF₃I
MW: 350.9 g/mol
Boiling Pt: 238.6 °C (760 mm Hg)
Melting Pt: 73…75 °C
Flash Pt: 98.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00672935
CAS Number: 364-12-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR