5-Bromo-2-iodobenzoic acid 98%
Supplier: Ambeed
|
Warning
|
A170429-100G
A170429-10G
A170429-500G
A170429-25G
A170429-5G
103844-770EA
6
USD
103844-770
76769-198
76769-200
76769-202
77081-880
77301-054
5-Bromo-2-iodobenzoic acid 98%
5-Bromo-2-iodobenzoic acid
Precaución: for Research Use Only
Formula:
C₇H₄BrIO₂ Boiling Pt: 360.1 °C (760 mm Hg) Melting Pt: 161…163 °C Flash Pt: 171.6 °C Storage Temperature: Ambient |
MDL Number:
MFCD00079717 CAS Number: 21740-00-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...