5-Bromo-2-iodobenzaldehyde 98%

Supplier: Ambeed
Warning

A111879-250MG A111879-1G A111879-25G
103797-916EA 12 USD
103797-916 103823-740 77108-022 77310-550 77684-992
5-Bromo-2-iodobenzaldehyde 98%
5-Bromo-2-iodobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄BrIO
MW: 310.9200 g/mol
Melting Pt: 89…90 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07779026
CAS Number: 689291-89-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR