5-Bromo-2-iodoaniline 98%
Supplier: Ambeed
|
Warning
|
A102577-100G
A102577-10G
A102577-250MG
A102577-25G
A102577-5G
A102577-1G
103796-390EA
7
USD
103796-390
103822-086
76809-340
76809-342
76809-344
76809-346
77282-948
77308-968
5-Bromo-2-iodoaniline 98%
5-Bromo-2-iodoaniline
Precaución: for Research Use Only
Formula:
C₆H₅BrIN MW: 297.9200 g/mol Boiling Pt: 309.5 °C (760 mm Hg) Flash Pt: 140.994 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD09753734 CAS Number: 64085-52-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...