(5-Bromo-2-pyridinyl)methanol 98%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-2-(hydroxymethyl)pyridine, 5-Bromo-2-hydroxymethylpyridine

A158161-100G A158161-10G A158161-250MG A158161-1G A158161-25G A158161-5G
103805-100EA 6 USD
103805-100 103843-308 77286-342 77286-344 77286-346 77319-156 77319-158 77319-160
(5-Bromo-2-pyridinyl)methanol 98%
(5-Bromo-2-pyridinyl)methanol

Precaución: for Research Use Only

Formula: C₆H₆BrNO
MW: 188.0200 g/mol
Melting Pt: 59…60 °C
Storage Temperature: Ambient
MDL Number: MFCD04035597
CAS Number: 88139-91-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR