5-Bromo-2-hydroxybenzonitrile 98%

Supplier: Ambeed
Warning

A164755-10G A164755-1G A164755-25G A164755-5G A164755-100G
103844-158EA 9 USD
103844-158 76768-762 76768-764 77320-344 77320-346 77625-620
5-Bromo-2-hydroxybenzonitrile 98%
5-Bromo-2-hydroxybenzonitrile

Precaución: for Research Use Only

Formula: C₇H₄BrNO
Melting Pt: 158…163 °C
Flash Pt: 124.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00143428
CAS Number: 40530-18-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR