5-Bromosalicylaldehyde 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-Bromo-2-hydroxybenzaldehyde
A218914-25G
A218914-1000G
A218914-100G
A218914-10G
A218914-500G
A218914-5G
103849-628EA
9
USD
103849-628
77088-168
77139-814
77139-816
77139-818
77139-820
77303-518
5-Bromosalicylaldehyde 98%
5-Bromosalicylaldehyde
Precaución: for Research Use Only
Formula:
C₇H₅BrO₂ Boiling Pt: 247.3 °C (760 mm Hg) Melting Pt: 102…106 °C Flash Pt: 103.4 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003330 CAS Number: 1761-61-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...