5-Bromo-2-hydroxy-4-picoline 95%

Supplier: Ambeed
Warning

A510668-1G A510668-5G A510668-100G A510668-10G A510668-500G
103828-390EA 11 USD
103828-390 103839-492 76843-476 76843-478 77673-348 77673-350 77882-259
5-Bromo-2-hydroxy-4-picoline 95%
5-Bromo-2-hydroxy-4-picoline

Precaución: for Research Use Only

Formula: C₆H₆BrNO
Boiling Pt: 291.8 °C (760 mm Hg)
Melting Pt: 198…202 °C
Flash Pt: 130.3 °C
Storage Temperature: Ambient
MDL Number: MFCD03427664
CAS Number: 164513-38-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR