5-Bromo-2-hydroxy-4-methoxybenzaldehyde 95%

Supplier: Ambeed
Warning

A536723-5G A536723-100MG A536723-250MG A536723-1G
103828-598EA 55 USD
103828-598 103839-720 76843-750 77157-028 77157-030 77307-760
5-Bromo-2-hydroxy-4-methoxybenzaldehyde 95%
5-Bromo-2-hydroxy-4-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₇BrO₃
MW: 231.043 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00839421
CAS Number: 57543-36-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR