5-Bromo-2-fluorophenol 98%

Supplier: Ambeed
Warning

A181577-25G A181577-5G A181577-100G A181577-10G A181577-500G
103845-998EA 7 USD
103845-998 77083-530 77083-532 77287-244 77287-246 77287-248
5-Bromo-2-fluorophenol 98%
5-Bromo-2-fluorophenol

Precaución: for Research Use Only

Formula: C₆H₄BrFO
MW: 191 g/mol
Boiling Pt: 205.8 °C (760 mm Hg)
Melting Pt: 96…100 °C
Flash Pt: 91.7 °C
Storage Temperature: Ambient
MDL Number: MFCD07782066
CAS Number: 112204-58-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR