1-Bromo-4-fluoro-3-nitrobenzene 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Bromo-1-fluoro-2-nitrobenzene, 5-Bromo-2-fluoronitrobenzene
A100815-1000G
A100815-100G
A100815-10G
A100815-25G
A100815-500G
A100815-5G
103821-334EA
5
USD
103821-334
77282-564
77282-566
77282-568
77282-570
77282-572
77282-574
1-Bromo-4-fluoro-3-nitrobenzene 98%
1-Bromo-4-fluoro-3-nitrobenzene
Precaución: for Research Use Only
Formula:
C₆H₃BrFNO₂ MW: 220 g/mol Boiling Pt: 257.5 °C (760 mm Hg) Melting Pt: 18…19 °C Flash Pt: 110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00129165 CAS Number: 364-73-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...