5-Bromo-2-fluoronicotinic acid 98%
Supplier: Ambeed
|
Warning
|
A351848-100G
A351848-10G
A351848-250MG
A351848-25G
A351848-5G
A351848-1G
103826-054EA
8
USD
103826-054
103837-050
76788-398
76788-400
76788-402
76840-162
76840-164
77306-618
5-Bromo-2-fluoronicotinic acid 98%
5-Bromo-2-fluoronicotinic acid
Precaución: for Research Use Only
Formula:
C₆H₃BrFNO₂ Boiling Pt: 327.4 °C (760 mm Hg) Storage Temperature: Refrigerator |
MDL Number:
MFCD06797939 CAS Number: 29241-66-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...