5-Bromo-2-fluorobenzonitrile 98%
Supplier: Ambeed
|
Warning
|
A250285-5G
A250285-100G
A250285-10G
A250285-500G
A250285-25G
103851-758EA
5
USD
103851-758
77091-110
77113-436
77113-438
77113-440
77304-516
5-Bromo-2-fluorobenzonitrile 98%
5-Bromo-2-fluorobenzonitrile
Precaución: for Research Use Only
Formula:
C₇H₃BrFN MW: 200.01 g/mol Boiling Pt: 220.4 °C (760 mm Hg) Melting Pt: 76…81 °C Flash Pt: 111.2 °C Storage Temperature: Ambient |
MDL Number:
MFCD00143424 CAS Number: 179897-89-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...