5-Bromo-2-fluorobenzoic acid 98%

Supplier: Ambeed
Warning

A114691-100G A114691-10G A114691-25G A114691-500G A114691-1G
103824-292EA 7 USD
103824-292 77271-144 77271-146 77271-148 77271-150 77747-835
5-Bromo-2-fluorobenzoic acid 98%
5-Bromo-2-fluorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₄BrFO₂
Melting Pt: 141…145 °C
Storage Temperature: Ambient
MDL Number: MFCD00143423
CAS Number: 146328-85-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR