5-Bromo-2-fluoro-4-methylpyridine 97%

Supplier: Ambeed
Warning

A102567-100G A102567-25G A102567-500G A102567-1G A102567-5G A102567-10G
103796-388EA 9 USD
103796-388 103822-084 76765-254 76765-256 76765-258 77308-964 77308-966 77623-836
5-Bromo-2-fluoro-4-methylpyridine 97%
5-Bromo-2-fluoro-4-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₅BrFN
MW: 190.01 g/mol
Flash Pt: 80.2 °C
Storage Temperature: Ambient
MDL Number: MFCD09839213
CAS Number: 864830-16-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR