5-Bromo-2-fluoro-4-methylbenzonitrile 98%
Supplier: Ambeed
|
Warning
|
A103869-10G
A103869-250MG
A103869-25G
A103869-1G
A103869-100G
103796-620EA
8
USD
103796-620
103822-330
76821-374
76821-376
76821-378
76821-380
77309-198
77884-945
5-Bromo-2-fluoro-4-methylbenzonitrile 98%
5-Bromo-2-fluoro-4-methylbenzonitrile
Precaución: for Research Use Only
Formula:
C₈H₅BrFN Storage Temperature: Ambient |
MDL Number:
MFCD18452117 CAS Number: 1269493-45-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...