5-Bromo-2-fluoro-4-methylaniline 98%

Supplier: Ambeed
Warning

A101643-10G A101643-250MG A101643-25G A101643-1G A101643-5G
103796-058EA 19 USD
103796-058 103821-754 76869-174 76869-176 76869-178 77297-180 77297-182
5-Bromo-2-fluoro-4-methylaniline 98%
5-Bromo-2-fluoro-4-methylaniline

Precaución: for Research Use Only

Formula: C₇H₇BrFN
MW: 204.0400 g/mol
Boiling Pt: 244 °C (760 mm Hg)
Flash Pt: 101 °C
Storage Temperature: Ambient
MDL Number: MFCD11504886
CAS Number: 945244-29-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR