5-Bromo-2-fluoro-m-xylene 98%

Supplier: Ambeed
Warning

A220015-100G A220015-10G A220015-1G A220015-25G A220015-5G
103818-832EA 5 USD
103818-832 103849-714 76770-788 76770-790 77088-300 77088-302 77088-304
5-Bromo-2-fluoro-m-xylene 98%
5-Bromo-2-fluoro-m-xylene

Precaución: for Research Use Only

Formula: C₈H₈BrF
MW: 203.05 g/mol
Boiling Pt: 205 °C (760 mm Hg)
Flash Pt: 80.4 °C
Storage Temperature: Ambient
MDL Number: MFCD01320701
CAS Number: 99725-44-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR