5-Bromo-2-ethylpyridine 98%

Supplier: Ambeed
Warning

A163388-100MG A163388-250MG A163388-1G A163388-5G
103805-722EA 36 USD
103805-722 103843-982 76783-968 76783-970 77320-076 77320-078
5-Bromo-2-ethylpyridine 98%
5-Bromo-2-ethylpyridine

Precaución: for Research Use Only

Formula: C₇H₈BrN
Boiling Pt: 201.1 °C (760 mm Hg)
Flash Pt: 75.436 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11226235
CAS Number: 38749-90-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR