5-Bromo-2-ethoxypyrimidine 98%
Supplier: Ambeed
|
Warning
|
A150498-10G
A150498-1G
A150498-25G
A150498-5G
103804-174EA
16
USD
103804-174
103842-308
77010-732
77299-808
77299-810
77299-812
5-Bromo-2-ethoxypyrimidine 98%
5-Bromo-2-ethoxypyrimidine
Precaución: for Research Use Only
Formula:
C₆H₇BrN₂O MW: 203.0400 g/mol Boiling Pt: 270.3 °C (760 mm Hg) Flash Pt: 117.3 °C Storage Temperature: Ambient |
MDL Number:
MFCD07375135 CAS Number: 17758-11-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...