5-Bromo-2-ethoxypyridine 98%

Supplier: Ambeed
Warning

A316998-25G A316998-5G A316998-1000G A316998-100G A316998-10G A316998-500G
103825-124EA 7 USD
103825-124 103836-056 76838-904 76838-906 77277-648 77277-650 77277-652 77277-654
5-Bromo-2-ethoxypyridine 98%
5-Bromo-2-ethoxypyridine

Precaución: for Research Use Only

Formula: C₇H₈BrNO
MW: 202.05 g/mol
Boiling Pt: 216.3 °C (760 mm Hg)
Melting Pt: 32…36 °C
Flash Pt: 103 °C
Storage Temperature: Ambient
MDL Number: MFCD00234311
CAS Number: 55849-30-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR