5-Bromo-2-cyclopropylpyridine 98%

Supplier: Ambeed
Warning

A115259-100MG A115259-250MG
103798-534EA 65 USD
103798-534 103824-388 77026-046 77026-048
5-Bromo-2-cyclopropylpyridine 98%
5-Bromo-2-cyclopropylpyridine

Precaución: for Research Use Only

Formula: C₈H₈BrN
MW: 198.06 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12033369
CAS Number: 579475-29-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR