5-Bromo-2-chloropyrimidine 98%

Supplier: Ambeed
Warning

A107210-100G A107210-10G A107210-1G A107210-25G A107210-500G A107210-5G
103822-916EA 6 USD
103822-916 76809-726 76809-728 76809-730 76809-732 76809-734 77309-770
5-Bromo-2-chloropyrimidine 98%
5-Bromo-2-chloropyrimidine

Precaución: for Research Use Only

Formula: C₄H₂BrClN₂
MW: 193.43 g/mol
Boiling Pt: 291 °C (760 mm Hg)
Melting Pt: 76…81 °C
Storage Temperature: Ambient
MDL Number: MFCD00483232
CAS Number: 32779-36-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR