5-Bromo-2-chloropyridine 98%

Supplier: Ambeed
Warning

A160865-100G A160865-10G A160865-1G A160865-500G A160865-25G A160865-5G
103843-662EA 6 USD
103843-662 76973-594 76973-596 76973-598 76973-600 77319-618 77319-620
5-Bromo-2-chloropyridine 98%
5-Bromo-2-chloropyridine

Precaución: for Research Use Only

Formula: C₅H₃BrClN
MW: 192.44 g/mol
Boiling Pt: 208.1 °C (760 mm Hg)
Melting Pt: 65…69 °C
Flash Pt: 79.7 °C
Storage Temperature: Ambient
MDL Number: MFCD01318951
CAS Number: 53939-30-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR