1-Bromo-4-chloro-3-nitrobenzene 98%

Supplier: Ambeed
Warning

A337999-100G A337999-10G A337999-1G A337999-25G A337999-5G
103825-762EA 5 USD
103825-762 103836-732 76773-116 76773-118 76839-752 76839-754 76839-756
1-Bromo-4-chloro-3-nitrobenzene 98%
1-Bromo-4-chloro-3-nitrobenzene

Precaución: for Research Use Only

Formula: C₆H₃BrClNO₂
Boiling Pt: 279.2 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD05863216
CAS Number: 16588-24-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR