5-Bromo-2-chloroisonicotinic acid 97%

Supplier: Ambeed
Warning

A130671-100G A130671-1G A130671-25G A130671-5G A130671-500G
103801-454EA 6 USD
103801-454 77009-910 77298-422 77313-904 77313-906 77683-161
5-Bromo-2-chloroisonicotinic acid 97%
5-Bromo-2-chloroisonicotinic acid

Precaución: For research use only.

Formula: C₆H₃BrClNO₂
MW: 236.4500 g/mol
Boiling Pt: 429.1 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD07375114
CAS Number: 886365-31-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR