5-Bromo-2-chlorobenzoic acid 98%

Supplier: Ambeed
Warning

A203856-100G A203856-10G A203856-500G A203856-25G
103809-390EA 88 USD
103809-390 76770-238 76770-240 76770-242 77302-876
5-Bromo-2-chlorobenzoic acid 98%
5-Bromo-2-chlorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₄BrClO₂
MW: 235.46 g/mol
Boiling Pt: 324.5 °C (760 mm Hg)
Melting Pt: 154…156 °C
Flash Pt: 150.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00002415
CAS Number: 21739-92-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR