5-Bromo-2-chloroanisole 98%

Supplier: Ambeed
Warning

A242848-25G A242848-5G A242848-100G A242848-10G A242848-500G
103811-490EA 5 USD
103811-490 103851-310 77090-498 77090-500 77098-506 77098-508 77098-510
5-Bromo-2-chloroanisole 98%
5-Bromo-2-chloroanisole

Precaución: for Research Use Only

Formula: C₇H₆BrClO
MW: 221.48 g/mol
Boiling Pt: 231.9 °C (760 mm Hg)
Melting Pt: 27…28 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00078659
CAS Number: 16817-43-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR