5-Bromo-2-chloroaniline 98%
Supplier: Ambeed
|
Warning
|
A187064-10G
A187064-1G
A187064-25G
A187064-5G
A187064-100G
103846-576EA
6
USD
103846-576
76813-770
76813-772
77084-268
77084-270
77611-814
5-Bromo-2-chloroaniline 98%
5-Bromo-2-chloroaniline
Precaución: for Research Use Only
Formula:
C₆H₅BrClN MW: 206.47 g/mol Boiling Pt: 255.8 °C (760 mm Hg) Melting Pt: 48 °C Flash Pt: 108.5 °C Storage Temperature: Ambient |
MDL Number:
MFCD06796283 CAS Number: 60811-17-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...