5-Bromo-2-chloro-7H-pyrrolo[2,3-d]pyrimidine 97%

Supplier: Ambeed
Warning

A139013-10G A139013-250MG A139013-1G A139013-25G A139013-5G
103802-686EA 39 USD
103802-686 76782-834 76782-836 77315-756 77315-758 77315-760
5-Bromo-2-chloro-7H-pyrrolo[2,3-d]pyrimidine 97%
5-Bromo-2-chloro-7H-pyrrolo[2,3-d]pyrimidine

Precaución: for Research Use Only

Formula: C₆H₃BrClN₃
MW: 232.4700 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09909707
CAS Number: 1060816-58-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR