5-Bromo-2-chloro-4-methylpyrimidine 97%

Supplier: Ambeed
Warning

A316150-100G A316150-10G A316150-250MG A316150-1G A316150-25G A316150-5G A316150-500G
103825-090EA 7 USD
103825-090 103836-016 76787-994 76787-996 76787-998 76838-854 76838-856 76838-858 77872-444
5-Bromo-2-chloro-4-methylpyrimidine 97%
5-Bromo-2-chloro-4-methylpyrimidine

Precaución: for Research Use Only

Formula: C₅H₄BrClN₂
Storage Temperature: Refrigerator
MDL Number: MFCD13193654
CAS Number: 633328-95-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR