5-Bromo-2-chloro-4-methyl-3-nitropyridine 95%
Supplier: Ambeed
|
Warning
|
A234490-1G
A234490-100G
A234490-10G
A234490-250MG
A234490-25G
A234490-5G
103810-952EA
7
USD
103810-952
103850-710
77089-696
77140-134
77140-136
77140-138
77303-978
77303-980
5-Bromo-2-chloro-4-methyl-3-nitropyridine 95%
5-Bromo-2-chloro-4-methyl-3-nitropyridine
Precaución: for Research Use Only
Formula:
C₆H₄BrClN₂O₂ MW: 251.47 Boiling Pt: 304.4 °C (760 mm Hg) Storage Temperature: Refrigerator |
MDL Number:
MFCD06659505 CAS Number: 884495-15-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...