5-Bromo-2-chloro-4-methoxypyridine 95%
Supplier: Ambeed
|
Warning
|
A116270-100MG
A116270-10G
A116270-250MG
A116270-25G
A116270-1G
A116270-5G
103798-702EA
54
USD
103798-702
103824-564
76646-860
76646-862
76646-864
76646-866
77297-442
77297-444
5-Bromo-2-chloro-4-methoxypyridine 95%
5-Bromo-2-chloro-4-methoxypyridine
Precaución: for Research Use Only
Formula:
C₆H₅BrClNO MW: 222.46 g/mol Boiling Pt: 251 °C (760 mm Hg) Flash Pt: 105 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD09909843 CAS Number: 880870-13-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...