5-Bromo-2-chloro-4-fluoropyridine 95%

Supplier: Ambeed
Warning

A178179-1G A178179-100MG
103807-302EA 319 USD
103807-302 103845-642 77301-546 77671-522
5-Bromo-2-chloro-4-fluoropyridine 95%
5-Bromo-2-chloro-4-fluoropyridine

Precaución: for Research Use Only

Formula: C₅H₂BrClFN
Storage Temperature: Refrigerator
MDL Number: MFCD09910137
CAS Number: 1211580-49-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR