5-Bromo-2-chloro-4-fluoroanisole 98%

Supplier: Ambeed
Warning

Synonyms: 1-Bromo-4-chloro-2-fluoro-5-methoxybenzene

A466471-1G A466471-5G A466471-100MG A466471-250MG A466471-25G
103827-980EA 16 USD
103827-980 103839-054 76842-964 76842-966 77156-266 77156-268 77307-448
5-Bromo-2-chloro-4-fluoroanisole 98%
5-Bromo-2-chloro-4-fluoroanisole

Precaución: for Research Use Only

Formula: C₇H₅BrClFO
MW: 239.47 g/mol
Boiling Pt: 237.7 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD07784265
CAS Number: 146447-18-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR