5-Bromo-2-chloro-4-(trifluoromethyl)pyridine 95%

Supplier: Ambeed
Warning

A164684-250MG A164684-1G
103805-890EA 24 USD
103805-890 103844-148 76997-972 77320-336
5-Bromo-2-chloro-4-(trifluoromethyl)pyridine 95%
5-Bromo-2-chloro-4-(trifluoromethyl)pyridine

Precaución: for Research Use Only

Formula: C₆H₂BrClF₃N
MW: 260.44 g/mol
Boiling Pt: 212.7 °C (760 mm Hg)
Flash Pt: 82.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11100520
CAS Number: 823221-93-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR