5-Bromo-2-chloro-3-methylpyridine 98%

Supplier: Ambeed
Warning

A101296-100G A101296-10G A101296-25G A101296-1G A101296-5G
103795-896EA 7 USD
103795-896 103821-582 77093-504 77093-506 77093-508 77297-046 77297-048
5-Bromo-2-chloro-3-methylpyridine 98%
5-Bromo-2-chloro-3-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₅BrClN
Boiling Pt: 234.2 °C (760 mm Hg)
Melting Pt: 40…44 °C
Flash Pt: 95.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03095093
CAS Number: 29241-60-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR